EMAIL THIS PAGE TO A FRIEND
17878 Sigma-Aldrich

Bromoform

puriss., ≥99.0% (GC)

Synonym: Tribromomethane

  • CAS Number 75-25-2

  • Empirical Formula (Hill Notation) CHBr3

  • Molecular Weight 252.73

  •  Beilstein Registry Number 1731048

  •  EC Number 200-854-6

  •  MDL number MFCD00000128

  •  PubChem Substance ID 329751575

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   8.7 (vs air)
vapor pressure   5 mmHg ( 20 °C)
InChI Key   DIKBFYAXUHHXCS-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
contains   ~0.05% amylene as stabilizer
refractive index   n20/D 1.595(lit.)
  n20/D 1.596
bp   146-150 °C(lit.)
mp   5-8 °C(lit.)
solubility   H2O: soluble
  acetone: miscible
  alcohol: miscible
  benzene: miscible
  chloroform: miscible
  diethyl ether: miscible
density   2.89 g/mL at 25 °C(lit.)

Description

Application

Bromoform was used to carry out the risk assessment of tribromomethane in drinking water.

General description

Polar ordering induced by pressure in bromoform has been determined using single-crystal X-ray diffraction.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2515 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

241032 Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?