178918 Sigma-Aldrich

Hexane, mixture of isomers

ACS reagent, ≥98.5%

DOWNLOAD MSDS (PDF)

Synonym: Hexanes, Isohexane

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ACS reagent
vapor density   3 (vs air)
description   Mixture of hexane isomers plus methylcyclopentane
assay   ≥98.5%
autoignition temp.   437 °F
expl. lim.   7.5 %
impurities   thiophene, passes test
  ≤0.0003 meq/g water-soluble titr. acid
  ≤0.005% S compounds
evapn. residue   ≤0.0010%
color   APHA: ≤10
refractive index   n20/D 1.379(lit.)
bp   68-70 °C(lit.)
mp   −95 °C(lit.)
density   0.672 g/mL at 25 °C(lit.)

Description

Packaging

1, 6×1, 2, 2.5, 4, 4×4 L in glass bottle

20 L in steel drum

200 L in Pure-Pac™ 2

200 L in steel drum

500, 6×500 mL in glass bottle

Price and Availability

Customers Also Viewed

Acetone
179124Acetone

ACS reagent, ≥99.5%

Dichloromethane

contains amylene as stabilizer, ACS reagent, ≥99.5%

Ethyl acetate

ACS reagent, ≥99.5%

Hexane
296090Hexane

anhydrous, 95%

Hexane, mixture of isomers

anhydrous, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?