EMAIL THIS PAGE TO A FRIEND
17892 Sigma-Aldrich

Sulfate-ionophore I

Selectophore, function tested

Synonym: 1,3-[Bis(3-phenylthioureidomethyl)]benzene

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   AKJHZJCPXMMQCY-UHFFFAOYSA-N
mp   178-182 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Neutral sulfate-selective ionophore for an electrode with remarkable sulfate selectivity

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?