• USA Home
  • 17900 - Bromopentafluorobenzene

EMAIL THIS PAGE TO A FRIEND
17900 Sigma-Aldrich

Bromopentafluorobenzene

purum, ≥97.0% (GC)

  • CAS Number 344-04-7

  • Linear Formula BrC6F5

  • Molecular Weight 246.96

  •  Beilstein Registry Number 1819390

  •  EC Number 206-449-0

  •  MDL number MFCD00000287

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C6, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   XEKTVXADUPBFOA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.449(lit.)
  n20/D 1.449
bp   137 °C(lit.)
mp   −31 °C(lit.)
density   1.981 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
CY9040552
Flash Point(F) 
189 °F
Flash Point(C) 
87 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B75158 Bromopentafluorobenzene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?