EMAIL THIS PAGE TO A FRIEND
17920 Sigma-Aldrich

1-Bromopentane

purum, ≥98.5% (GC)

Synonym: n-Amyl bromide, Pentyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   >1 (vs air)
InChI Key   YZWKKMVJZFACSU-UHFFFAOYSA-N
grade   purum
assay   ≥98.5% (GC)
refractive index   n20/D 1.444(lit.)
  n20/D 1.445
bp   130 °C(lit.)
mp   −95 °C(lit.)
density   1.218 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
RZ9770000
Flash Point(F) 
89.6 °F
Flash Point(C) 
32 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?