179418 Sigma-Aldrich

Toluene

ACS reagent, ≥99.5%

DOWNLOAD MSDS (PDF)

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ACS reagent
vapor density   3.2 (vs air)
vapor pressure   22 mmHg ( 20 °C)
  26 mmHg ( 25 °C)
assay   ≥99.5%
autoignition temp.   997 °F
expl. lim.   7 %
impurities   H2SO4, passes test(darkened)
  ≤0.003% S compounds
  ≤0.030% water
evapn. residue   ≤0.0010%
color   APHA: ≤10
refractive index   n/D 1.496(lit.)
bp   110-111 °C(lit.)
mp   -93 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)

Description

Packaging

1, 6×1, 2, 2.5, 4×2.5, 4, 4×4 L in glass bottle

20, 200 L in steel drum

View returnable container options.

400 L in Kilo-Lab™

500, 6×500 mL in glass bottle

Price and Availability

Customers Also Viewed

Toluene
244511Toluene

anhydrous, 99.8%

Toluene
650579Toluene

CHROMASOLV® Plus, for HPLC, ≥99.9%

Toluene
155004Toluene

ACS spectrophotometric grade, ≥99.5%

Toluene
320552Toluene

ACS reagent, ≥99.5%

Toluene
676756Toluene

ACS reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?