• USA Home
  • 17947 - Maize powder MA 1, IRMM certified reference material No. 412

EMAIL THIS PAGE TO A FRIEND
17947 Sigma-Aldrich

Maize powder MA 1, IRMM certified reference material No. 412

BioChemika, for the detection of GMO maize (Bt-11) with PCR

Synonym: GMO maize powder MA 1

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, Reference Materials from the European Commission′s Joint Research Centre (formerly IRMM)
product line   BioChemika
CofA   certificate is enclosed in each package
quality   for the detection of GMO maize (Bt-11) with PCR
composition   genetically modified Bt-11 maize, 1%
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   matrix material

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?