• USA Home
  • 17968 - Fluorescent IEF-Marker pI 10.3

EMAIL THIS PAGE TO A FRIEND
17968 Sigma-Aldrich

Fluorescent IEF-Marker pI 10.3

Synonym: IEF-Marker fluorescent pI 10.3

Purchase

Description

Application

suitable as IEF-Marker for HPCE and for IEF-gels

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Reconstitution

Dissolve the IEF-marker at 1mg/ml in H2O or 10 mM HCl, sonicate for up to 10 min if necessary. Store at 4 °C (the solution is stable for approximately 6 months). For use in HPCE this stock solution should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the 1 mg/ml stock solution can be loaded directly onto the gel (max. 1 μl per lane).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

77672 Fluorescent IEF-Marker pI 10.3 solution, 1 mg/mL in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?