• USA Home
  • 17970 - 2,4′-Dibromoacetophenone

EMAIL THIS PAGE TO A FRIEND
17970 Sigma-Aldrich

2,4′-Dibromoacetophenone

puriss., ≥98.5% (HPLC)

Synonym: 4′-Bromophenacyl bromide

  • CAS Number 99-73-0

  • Linear Formula BrC6H4COCH2Br

  • Molecular Weight 277.94

  •  Beilstein Registry Number 607604

  •  EC Number 202-783-6

  •  MDL number MFCD00000200

  •  PubChem Substance ID 329751593

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   FKJSFKCZZIXQIP-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (HPLC)
mp   108-110 °C(lit.)
  108-112 °C
Gene Information   human ... PLA2G2A(5320)
rat ... Pla2g2a(29692)

Description

Other Notes

Modifies histidine residues in proteins

General description

The effect of 2,4′-dibromoacetophenone on rat seminiferous tubules in vitro was studied.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
2
RTECS 
AM6950000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D38308 2,4′-Dibromoacetophenone, >98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?