EMAIL THIS PAGE TO A FRIEND
18000 Sigma-Aldrich

3-Bromophenol

purum, ≥97.5% (HPLC)

  • CAS Number 591-20-8

  • Linear Formula BrC6H4OH

  • Molecular Weight 173.01

  •  Beilstein Registry Number 1853950

  •  EC Number 209-706-5

  •  MDL number MFCD00002253

  •  PubChem Substance ID 24850841

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   MNOJRWOWILAHAV-UHFFFAOYSA-N
grade   purum
assay   ≥97.5% (HPLC)
bp   236 °C(lit.)
mp   28-32 °C(lit.)
  28-32 °C
solubility   alcohol: soluble
  diethyl ether: soluble
Gene Information   human ... ALOX12(239), ALOX15(246)

Description

Caution

may form a subcooled melt

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

3-Bromophenol was used in the synthesis of 3-lithium-o-lithiophenoxide.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

101079 3-Bromophenol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?