• USA Home
  • 18068 - 3-Bromophenylhydrazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
18068 Sigma-Aldrich

3-Bromophenylhydrazine hydrochloride

technical, ≥96% (AT)

  • CAS Number 27246-81-7

  • Linear Formula BrC6H4NHNH2·HCl

  • Molecular Weight 223.50

  •  Beilstein Registry Number 3565829

  •  EC Number 248-362-0

  •  MDL number MFCD00012933

  •  PubChem Substance ID 24850886

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   RPYIPFXHIKXRKS-UHFFFAOYSA-N
grade   technical
assay   ≥96% (AT)
mp   227-231 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGII
WGK Germany 
2
RTECS 
MV0815000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?