• USA Home
  • 18080 - 3-Bromo-1-phenyl-1-propene

EMAIL THIS PAGE TO A FRIEND
18080 Sigma-Aldrich

3-Bromo-1-phenyl-1-propene

purum, ≥98.0% (GC)

Synonym: 3-Phenylallyl bromide, Cinnamyl bromide

Purchase

Properties

Related Categories Alkenyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   RUROFEVDCUGKHD-QPJJXVBHSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.621(lit.)
bp   103 °C/22 mmHg(lit.)
mp   26-29 °C(lit.)
  27-30 °C
density   1.332 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Application

3-Bromo-1-phenyl-1-propene was used in the synthesis of peracetylated 2(I)-O-, 3(I)-O- and 6(I)-O-alkylated α-cyclodextrin.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C81217 3-Bromo-1-phenyl-1-propene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?