EMAIL THIS PAGE TO A FRIEND
18103 Sigma-Aldrich

1-Bromopinacolone

purum, ≥97.0% (GC)

Synonym: 1-Bromo-3,3-dimethyl-2-butanone, Bromomethyl tert-butyl ketone

  • CAS Number 5469-26-1

  • Linear Formula (CH3)3CCOCH2Br

  • Molecular Weight 179.05

  •  Beilstein Registry Number 506485

  •  EC Number 226-794-0

  •  MDL number MFCD00000206

  •  PubChem Substance ID 24850907

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   SAIRZMWXVJEBMO-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.466(lit.)
  n20/D 1.466
bp   188-194 °C(lit.)
mp   −10 °C(lit.)
density   1.331 g/mL at 25 °C(lit.)
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
RTECS 
EL7010000
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

414131 1-Bromopinacolone, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?