• USA Home
  • 18167 - (S)-(−)-2-Bromopropionic acid

EMAIL THIS PAGE TO A FRIEND
18167 Sigma-Aldrich

(S)-(−)-2-Bromopropionic acid

purum, ≥98.5% (sum of enantiomers, GC)

  • CAS Number 32644-15-8

  • Linear Formula CH3CH(Br)CO2H

  • Molecular Weight 152.97

  •  Beilstein Registry Number 1720261

  •  MDL number MFCD00137325

  •  PubChem Substance ID 329751643

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   MONMFXREYOKQTI-REOHCLBHSA-N
grade   purum
assay   ≥98.5% (sum of enantiomers, GC)
optical activity   [α]20/D −26.5±2°, neat
refractive index   n20/D 1.475
bp   78 °C/4 mmHg(lit.)
mp   −35 °C(lit.)
density   1.696 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Caution

may discolor to brownish on storage

Other Notes

Chiral building block; synthesis of N-substituted D-alanines with amines; Preparation of chiral ethers and esters

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

385514 (S)-(−)-2-Bromopropionic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?