• USA Home
  • 18170 - (±)-2-Bromopropionic acid

EMAIL THIS PAGE TO A FRIEND
18170 Sigma-Aldrich

(±)-2-Bromopropionic acid

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   MONMFXREYOKQTI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.475(lit.)
  n20/D 1.475
bp   203 °C(lit.)
density   1.7 g/mL at 25 °C(lit.)

Description

Application

(±)-2-Bromopropionic acid was used in the synthesis of S-(1-carboxypropyl)-L-cysteine (1-CPC).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
UA2451715
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?