• USA Home
  • 18210 - 2-Bromopropionyl bromide

EMAIL THIS PAGE TO A FRIEND
18210 Sigma-Aldrich

2-Bromopropionyl bromide

purum, ≥97.0% (AT)

  • CAS Number 563-76-8

  • Linear Formula CH3CHBrCOBr

  • Molecular Weight 215.87

  •  Beilstein Registry Number 1071331

  •  EC Number 209-261-7

  •  MDL number MFCD00000142

  •  PubChem Substance ID 24850977

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor density   7.5 (vs air)
vapor pressure   1.3 mmHg ( 20 °C)
InChI Key   ILLHORFDXDLILE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
refractive index   n20/D 1.518(lit.)
  n20/D 1.520
bp   48-50 °C/10 mmHg(lit.)
density   2.03 g/mL at 20 °C
  2.061 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

2-Bromopropionyl bromide was used in the synthesis of ethyl cellulose macroinitiator by direct acylation of ethyl cellulose.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

249785 2-Bromopropionyl bromide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?