• USA Home
  • 18220 - Methyl 3-bromopropionate

EMAIL THIS PAGE TO A FRIEND
18220 Sigma-Aldrich

Methyl 3-bromopropionate

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   KQEVIFKPZOGBMZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.458(lit.)
  n20/D 1.459
bp   64-66 °C/18 mmHg(lit.)
density   1.53 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
UA2451790
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?