EMAIL THIS PAGE TO A FRIEND
18350 Sigma-Aldrich

N-Bromosuccinimide

purum, ≥95.0% (RT)

Synonym: NBS

  • CAS Number 128-08-5

  • Empirical Formula (Hill Notation) C4H4BrNO2

  • Molecular Weight 177.98

  •  Beilstein Registry Number 113916

  •  EC Number 204-877-2

  •  MDL number MFCD00005510

  •  PubChem Substance ID 24851059

Purchase

Properties

Related Categories Bromination, C-X Bond Formation (Halogen), Chemical Synthesis, Synthetic Reagents
InChI Key   PCLIMKBDDGJMGD-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (RT)
mp   175-180 °C (dec.)(lit.)
  175-180 °C (dec.)

Description

Application

Versatile brominating agent. For the oxidation of tryptophan though tyrosine, histidine and methionine residues may be oxidized to a lesser extent. Also used for the modification of ribosomal sulfhydryl groups.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

B81255 N-Bromosuccinimide, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?