EMAIL THIS PAGE TO A FRIEND
18422 Sigma-Aldrich

Glass wool

for laboratory use

Purchase

Properties

InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
impurities   ≤0.001% heavy metals (as Pb)
  ≤0.1% alkalis (as Na2O)
  ≤0.8% soluble in HCl
cation traces   Fe: ≤50 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
nwg
RTECS 
VV7310000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?