EMAIL THIS PAGE TO A FRIEND
18438 Sigma-Aldrich

Thionyl chloride

≥98%

Purchase

Properties

Related Categories C-X Bond Formation (Halogen), Chemical Synthesis, Chlorination, Synthetic Reagents
vapor pressure   97 mmHg ( 20 °C)
InChI Key   FYSNRJHAOHDILO-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.518(lit.)
bp   79 °C(lit.)
mp   −105 °C(lit.)
density   1.631 g/mL at 25 °C(lit.)

Description

General description

appearance: colorless-yellowish, clear liquid

Price and Availability


Palau Chlor
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with water liberates toxic gas., Reacts violently with water.
RIDADR 
UN 1836 8 / PGI
WGK Germany 
1
RTECS 
XM5150000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?