EMAIL THIS PAGE TO A FRIEND
18500 Sigma-Aldrich

4-Bromotoluene

puriss., ≥99.0% (GC)

  • CAS Number 106-38-7

  • Linear Formula CH3C6H4Br

  • Molecular Weight 171.03

  •  Beilstein Registry Number 1903636

  •  EC Number 203-391-8

  •  MDL number MFCD00000109

Purchase

Properties

InChI Key   ZBTMRBYMKUEVEU-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
bp   184 °C(lit.)
mp   26-28 °C
  26-29 °C(lit.)
density   1.39 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XS7965600
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

B82200 4-Bromotoluene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?