EMAIL THIS PAGE TO A FRIEND
18540 Sigma-Aldrich

1-Bromotridecane

technical, ≥98.0% (GC)

Synonym: Tridecyl bromide

  • CAS Number 765-09-3

  • Linear Formula CH3(CH2)12Br

  • Molecular Weight 263.26

  •  Beilstein Registry Number 1739992

  •  EC Number 212-138-0

  •  MDL number MFCD00000227

  •  PubChem Substance ID 329751686

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   9.2 (vs air)
InChI Key   BFDNZQUBFCYTIC-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (GC)
refractive index   n20/D 1.459(lit.)
  n20/D 1.459
bp   148-150 °C/10 mmHg(lit.)
mp   4-7 °C(lit.)
density   1.03 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YD3100000
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

192422 1-Bromotridecane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?