Synonym: Ethylene glycol monomethyl ether, Methyl glycol
CAS Number 109-86-4
Linear Formula CH3OCH2CH2OH
Molecular Weight 76.09
Beilstein Registry Number 1731074
EC Number 203-713-7
MDL number MFCD00002867
PubChem Substance ID 24851142
Popular Documents: Specification Sheet (PDF)
| Related Categories | ReagentPlus, ReagentPlus Solvent Grade Products, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Flex-Spout Cans, |
| grade | ReagentPlus® |
| vapor density | 2.62 (vs air) |
| vapor pressure | 6.17 mmHg ( 20 °C) |
| assay | ≥99.0% |
| autoignition temp. | 548 °F |
| contains | 50 ppm BHT as stabilizer |
| expl. lim. | 24.5 % |
20 L in steel drum
500 mL in glass bottle
4 L in glass bottle
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?