• USA Home
  • 18550 - 4′-Bromo-2,2,2-trifluoroacetophenone

EMAIL THIS PAGE TO A FRIEND
18550 Sigma-Aldrich

4′-Bromo-2,2,2-trifluoroacetophenone

purum, ≥98.0% (GC)

Synonym: 4-Bromo-α,α,α-trifluoroacetophenone

  • CAS Number 16184-89-7

  • Linear Formula BrC6H4COCF3

  • Molecular Weight 253.02

  •  Beilstein Registry Number 1956460

  •  MDL number MFCD00191862

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   IHGSAQHSAGRWNI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   95 °C/4 mmHg(lit.)
mp   26-30 °C(lit.)
  26-30 °C
density   1.662 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

403229 4′-Bromo-2,2,2-trifluoroacetophenone, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?