• USA Home
  • 18555 - 1-Bromo-4-(trimethylsiloxy)benzene

EMAIL THIS PAGE TO A FRIEND
18555 Sigma-Aldrich

1-Bromo-4-(trimethylsiloxy)benzene

purum, ≥97.0% (GC)

Synonym: (4-Bromophenoxy)trimethylsilane

Purchase

Properties

InChI Key   HWKPTBHJWBSOOP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.515(lit.)
  n20/D 1.515
density   1.268 g/mL at 25 °C(lit.)

Description

Other Notes

Protected 4-bromophenol; employed e.g. for preparing the corresponding Grignard reagent

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

348201 1-Bromo-4-(trimethylsiloxy)benzene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?