• USA Home
  • 18675 - Ethyl 5-bromovalerate

EMAIL THIS PAGE TO A FRIEND
18675 Sigma-Aldrich

Ethyl 5-bromovalerate

purum, ≥97.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   AFRWBGJRWRHQOV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.458(lit.)
  n20/D 1.459
bp   104-109 °C/12 mmHg(lit.)
density   1.305 g/mL at 20 °C
  1.321 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
219.2 °F
Flash Point(C) 
104 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

129100 Ethyl 5-bromovalerate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?