• USA Home
  • 18735 - Hydrobromic acid solution

EMAIL THIS PAGE TO A FRIEND
18735 Sigma-Aldrich

Hydrobromic acid solution

33 wt. % in acetic acid

Synonym: Hydrogen bromide in acetic acid

  • Empirical Formula (Hill Notation) HBr

  • Molecular Weight 80.91

  •  Beilstein Registry Number 3587158

  •  MDL number MFCD00011323

  •  PubChem Substance ID 24851269

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Inorganic Acids, Synthetic Reagents More...
assay   31.5-34.5% (T)
concentration   33 wt. % in acetic acid
impurities   ≤1% water (KFT)
density   1.40 g/mL at 20 °C

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

1 L in glass bottle

50, 250 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
1
Flash Point(F) 
>149 °F
Flash Point(C) 
>65 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?