EMAIL THIS PAGE TO A FRIEND
18881 Sigma-Aldrich

1,3-Butadiene

purum, ≥98.0%

  • CAS Number 106-99-0

  • Linear Formula CH2=CHCH=CH2

  • Molecular Weight 54.09

  •  Beilstein Registry Number 605258

  •  EC Number 203-450-8

  •  MDL number MFCD00008659

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   1.9 (15 °C, vs air)
vapor pressure   1863 mmHg ( 21 °C)
InChI Key   KAKZBPTYRLMSJV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0%
autoignition temp.   788 °F
expl. lim.   12 %
bp   −4.5 °C(lit.)
mp   −109 °C(lit.)
density   0.62 g/mL at 20 °C(lit.)

Description

Biochem/physiol Actions

Environmental carcinogen. Induces cardiac hemangiosarcomas in mice.

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Packaging

pressure tin with 250 mL (net ~155 g)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2037F 2.1
WGK Germany 
3
Flash Point(F) 
-103 °F
Flash Point(C) 
-75 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

control valve

Product #

Description

Add to Cart

83547 Regulation valve, suitable for 100 mL and 250 mL pressure tins

replaced by

Product #

Description

Add to Cart

295035 1,3-Butadiene, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?