EMAIL THIS PAGE TO A FRIEND
18940 Sigma-Aldrich

(±)-1,3-Butanediol

ReagentPlus®, ≥99.0%

Synonym: 1,3-Butylene glycol

  • CAS Number 107-88-0

  • Linear Formula CH3CH(OH)CH2CH2OH

  • Molecular Weight 90.12

  •  Beilstein Registry Number 1731276

  •  EC Number 203-529-7

  •  MDL number MFCD00004554

  •  eCl@ss 39020505

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
grade   ReagentPlus®
vapor density   3.1 (20 °C, vs air)
vapor pressure   0.06 mmHg ( 20 °C)
InChI Key   PUPZLCDOIYMWBV-UHFFFAOYSA-N
assay   ≥99.0%
autoignition temp.   741 °F
refractive index   n20/D 1.44(lit.)
  n20/D 1.440
bp   203-204 °C(lit.)
density   1.005 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
1
RTECS 
EK0440000
Flash Point(F) 
226.4 °F
Flash Point(C) 
108 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?