EMAIL THIS PAGE TO A FRIEND
18969 Sigma-Aldrich

meso-2,3-Butanediol

puriss., ≥99.0% (GC)

  • CAS Number 5341-95-7

  • Linear Formula CH3CH(OH)CH(OH)CH3

  • Molecular Weight 90.12

  •  Beilstein Registry Number 1718900

  •  MDL number MFCD00077669

  •  PubChem Substance ID 329751763

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   OWBTYPJTUOEWEK-ZXZARUISSA-N
grade   puriss.
assay   ≥99.0% (GC)
bp   183-184 °C(lit.)
mp   32-34 °C(lit.)
  32-36 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

361461 meso-2,3-Butanediol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?