EMAIL THIS PAGE TO A FRIEND
18980 Sigma-Aldrich

2,3-Butanediol

purum, mixture of racemic and meso forms, ≥98.0% (GC)

Synonym: 2,3-Butylene glycol

  • CAS Number 513-85-9

  • Linear Formula CH3CH(OH)CH(OH)CH3

  • Molecular Weight 90.12

  •  Beilstein Registry Number 969165

  •  EC Number 208-173-6

  •  MDL number MFCD00004523

  •  PubChem Substance ID 24851398

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   OWBTYPJTUOEWEK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.433(lit.)
  n20/D 1.438
bp   183-184 °C(lit.)
mp   25 °C(lit.)
density   1.002 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
EK0532000
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

B84904 2,3-Butanediol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?