• USA Home
  • 19027 - 1-Butanesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
19027 Sigma-Aldrich

1-Butanesulfonyl chloride

purum, ≥98.0% (GC)

  • CAS Number 2386-60-9

  • Linear Formula CH3(CH2)3SO2Cl

  • Molecular Weight 156.63

  •  Beilstein Registry Number 1748742

  •  EC Number 219-204-8

  •  MDL number MFCD00007464

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
vapor pressure   0.5 mmHg ( 20 °C)
InChI Key   WEDIIKBPDQQQJU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.454(lit.)
  n20/D 1.455
bp   80-81 °C/9 mmHg(lit.)
density   1.208 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
RTECS 
EK5950000
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

263605 1-Butanesulfonyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?