190764 Sigma-Aldrich

2-Propanol

ACS reagent, ≥99.5%

DOWNLOAD MSDS (PDF)

Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Properties

Related Categories 2-Propanol (Isopropanol), ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles,
Quality Level   PREMIUM
grade   ACS reagent
vapor density   2.1 (vs air)
vapor pressure   33 mmHg ( 20 °C)
  44 mmHg ( 25 °C)
assay   ≥99.5%
autoignition temp.   750 °F
expl. lim.   2 %
impurities   ≤0.0001 meq/g Titr. acid or base
  ≤0.002% carbonyl compounds (as propionaldehyde or acetone)
  ≤0.2% water
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.377(lit.)
bp   82 °C(lit.)
mp   −89.5 °C(lit.)
solubility   H2O: passes test
density   0.785 g/mL at 25 °C(lit.)

Description

Packaging

1, 6×1, 2, 2.5, 4, 4×4 L in glass bottle

4×4 L in poly bottle

View returnable container options.

20, 200 L in steel drum

500, 6×500 mL in glass bottle

Price and Availability

Customers Also Viewed

2-Propanol

BioReagent, for molecular biology, ≥99%

2-Propanol

anhydrous, 99.5%

2-Propanol

ACS reagent, ≥99.5%

2-Propanol

ACS reagent, ≥99.5%

Ethanol
459844Ethanol

ACS reagent, ≥99.5% (200 proof), absolute

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?