• USA Home
  • 19201 - (S)-(−)-3-Butyn-2-ol

EMAIL THIS PAGE TO A FRIEND
19201 Sigma-Aldrich

(S)-(−)-3-Butyn-2-ol

purum, ≥98.0% (GC)

  • CAS Number 2914-69-4

  • Linear Formula HC≡CCH(OH)CH3

  • Molecular Weight 70.09

  •  Beilstein Registry Number 4652597

  •  MDL number MFCD00190166

  •  PubChem Substance ID 329751833

Purchase

Properties

InChI Key   GKPOMITUDGXOSB-BYPYZUCNSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.426(lit.)
bp   108-111 °C(lit.)
density   0.883 g/mL at 20 °C(lit.)

Description

Other Notes

Chiral building block

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2929 3(6.1) / PGII
WGK Germany 
3
Flash Point(F) 
78.8 °F
Flash Point(C) 
26 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

464007 (S)-(−)-3-Butyn-2-ol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?