EMAIL THIS PAGE TO A FRIEND
19364 Sigma-Aldrich

Sodium butyrate

≥99.0% (GC)

Synonym: Butyric acid sodium salt

  • CAS Number 156-54-7

  • Linear Formula CH3CH2CH2COONa

  • Molecular Weight 110.09

  •  Beilstein Registry Number 3629439

  •  EC Number 205-857-6

  •  MDL number MFCD00002816

  •  PubChem Substance ID 329751856

Purchase

Properties

InChI Key   MFBOGIVSZKQAPD-UHFFFAOYSA-M
assay   ≥99.0% (GC)
impurities   ≤4% water
mp   250-253 °C(lit.)

Description

Other Notes

Agent which alters chromatin structure; Enhanced transfection efficiency and improved cell survival after electroporation of G2/M-synchronized cells and treatment with sodium butyrate.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
ET6400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

B5887 Sodium butyrate, ≥98.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?