• USA Home
  • 19405 - 3,3-Dimethylbutyryl chloride

EMAIL THIS PAGE TO A FRIEND
19405 Sigma-Aldrich

3,3-Dimethylbutyryl chloride

puriss., ≥98.5% (GC)

Synonym: tert-Butylacetyl chloride

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   BUTKIHRNYUEGKB-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
refractive index   n20/D 1.421(lit.)
bp   127-129 °C(lit.)
density   0.969 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B88802 3,3-Dimethylbutyryl chloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?