• USA Home
  • 19515 - L-Valine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
19515 Sigma-Aldrich

L-Valine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

  • CAS Number 17498-50-9

  • Empirical Formula (Hill Notation) C5H11NO2 · HCl

  • Molecular Weight 153.61

  •  Beilstein Registry Number 4149740

  •  MDL number MFCD03791069

  •  PubChem Substance ID 329751892

  •  eCl@ss 32160406

Purchase

Properties

Related Categories Amino Acid Standards, Amino Acid, Peptide & Protein Standards, Analytical Standards, Analytical/Chromatography
grade   analytical standard
InChI Key   JETBVOLWZWPMKR-WCCKRBBISA-N
concentration   100 mM amino acid in 0.1 M HCl
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

General description

L-Valine hydrochloride is a semiorganic nonlinear optical material.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?