EMAIL THIS PAGE TO A FRIEND
19690 Sigma-Aldrich

2-Bromobutane

purum, ≥98.0% (GC)

Synonym: sec-Butyl bromide

  • CAS Number 78-76-2

  • Linear Formula CH3CH2CHBrCH3

  • Molecular Weight 137.02

  •  Beilstein Registry Number 505949

  •  EC Number 201-140-7

  •  MDL number MFCD00000156

  •  PubChem Substance ID 329751932

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   UPSXAPQYNGXVBF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.437(lit.)
  n20/D 1.437
bp   91 °C(lit.)
density   1.255 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2339 3 / PGII
WGK Germany 
2
RTECS 
EJ6228000
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles

Articles

GC Analysis of 2-Halohydrocarbon Enantiomers on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of 2-Halohydrocarbon Enantiomers on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B59500 2-Bromobutane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?