• USA Home
  • 19700 - 2-Bromo-2-methylpropane

EMAIL THIS PAGE TO A FRIEND
19700 Sigma-Aldrich

2-Bromo-2-methylpropane

purum, ≥97.0% (GC)

Synonym: 2-methyl-2-bromopropane, tert-Butyl bromide

  • CAS Number 507-19-7

  • Linear Formula (CH3)3CBr

  • Molecular Weight 137.02

  •  Beilstein Registry Number 1730892

  •  EC Number 208-065-9

  •  MDL number MFCD00000125

  •  PubChem Substance ID 24851816

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   RKSOPLXZQNSWAS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.4279(lit.)
  n20/D 1.428
bp   71-73 °C(lit.)
mp   −20 °C(lit.)
density   1.22 g/mL at 20 °C(lit.)

Description

Caution

may discolor to brownish-yellow on storage

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2342 3 / PGII
WGK Germany 
3
RTECS 
TX4150000
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

135615 2-Bromo-2-methylpropane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?