EMAIL THIS PAGE TO A FRIEND
19740 Sigma-Aldrich

tert-Butyl carbazate

purum, ≥98.0% (GC)

Synonym: tert-Butoxycarbonyl hydrazide, Boc-hydrazide

  • CAS Number 870-46-2

  • Linear Formula (CH3)3COCONHNH2

  • Molecular Weight 132.16

  •  Beilstein Registry Number 1756967

  •  EC Number 212-795-3

  •  MDL number MFCD00007593

  •  PubChem Substance ID 329751941

Purchase

Properties

Related Categories Amine Protection, Chemical Biology, Chemical Synthesis, Other Protecting and Derivatizing Reagents, Peptide Chemistry,
InChI Key   DKACXUFSLUYRFU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   63-65 °C/0.1 mmHg(lit.)
mp   38-42 °C
  39-42 °C(lit.)

Description

Other Notes

Starting material for preparing BOC-azide, a reagent to introduce the BOC amino protection; E. Wünsch in Houben-Weyl, Vol. 15/1, Synthese von grossen Peptiden, p. 112.; Reagent for the synthesis of sulfonic and carboxylic hydrazides

This product has been replaced by B91005-ALDRICH | tert-Butyl carbazate 98%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?