• USA Home
  • 19810 - tert-Butylcyclohexane

EMAIL THIS PAGE TO A FRIEND
19810 Sigma-Aldrich

tert-Butylcyclohexane

purum, ≥97.0% (GC)

Synonym: 2-Cyclohexyl-2-methylpropane

  • CAS Number 3178-22-1

  • Linear Formula (C6H11)C(CH3)3

  • Molecular Weight 140.27

  •  Beilstein Registry Number 1900578

  •  EC Number 221-652-4

  •  MDL number MFCD00001471

Purchase

Properties

vapor pressure   5 mmHg ( 37.7 °C)
InChI Key   XTVMZZBLCLWBPM-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.447(lit.)
  n20/D 1.447
bp   167 °C(lit.)
density   0.831 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3295C 3 / PGIII
WGK Germany 
3
RTECS 
GU9384375
Flash Point(F) 
107.6 °F
Flash Point(C) 
42 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?