• USA Home
  • 19820 - 4-tert-Butylcyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
19820 Sigma-Aldrich

4-tert-Butylcyclohexanecarboxylic acid

purum, ≥98.0% (T)

  • CAS Number 5451-55-8

  • Linear Formula (CH3)3CC6H10CO2H

  • Molecular Weight 184.28

  •  Beilstein Registry Number 2043187

  •  MDL number MFCD00042622

  •  PubChem Substance ID 329751949

Purchase

Properties

InChI Key   QVQKEGYITJBHRQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   148 °C(lit.)
  95-98 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

374938 4-tert-Butylcyclohexanecarboxylic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?