EMAIL THIS PAGE TO A FRIEND
19930 Sigma-Aldrich

1-Butene oxide

purum, ≥99.0% (GC)

Synonym: 1,2-Butylene oxide, Ethyloxirane

  • CAS Number 106-88-7

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 102411

  •  EC Number 203-438-2

  •  MDL number MFCD00005153

  •  PubChem Substance ID 329751956

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   RBACIKXCRWGCBB-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.384
bp   61-65 °C(lit.)
density   0.829 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3022 3 / PGII
WGK Germany 
2
RTECS 
EK3675000
Flash Point(F) 
5 °F
Flash Point(C) 
-15 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?