• USA Home
  • 19978 - tert-Butylhydrazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
19978 Sigma-Aldrich

tert-Butylhydrazine hydrochloride

purum, ≥97.0% (AT)

  • CAS Number 7400-27-3

  • Linear Formula (CH3)3CNHNH2 · HCl

  • Molecular Weight 124.61

  •  Beilstein Registry Number 3651224

  •  EC Number 231-001-6

  •  MDL number MFCD00012947

  •  PubChem Substance ID 329751962

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   DDPWVABNMBRBFI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   191-194 °C(lit.)
solubility   H2O: soluble 1 g/10 mL clear, colorless

Description

Application

tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines R-NH-N(PPh2)2 (where R = tBu).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LU4220000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?