EMAIL THIS PAGE TO A FRIEND
201812 Sigma-Aldrich

Acid Orange 74

Dye content 60 %

  • CAS Number 10127-27-2

  • Empirical Formula (Hill Notation) C16H11CrN5NaO8S

  • Molecular Weight 508.34

  •  Colour Index Number 18745

  •  EC Number 233-357-8

  •  MDL number MFCD08461055

  •  PubChem Substance ID 24852023

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   DFUHZEIMQHFCKR-GLCFPVLVSA-J
composition   Dye content, 60%
absorption   λmax 455 nm

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?