• USA Home
  • 20198 - tert-Butylmagnesium chloride solution

EMAIL THIS PAGE TO A FRIEND
20198 Sigma-Aldrich

tert-Butylmagnesium chloride solution

technical, ~1 M in THF

  • CAS Number 677-22-5

  • Linear Formula (CH3)3CMgCl

  • Molecular Weight 116.87

  •  Beilstein Registry Number 3535403

  •  MDL number MFCD00000465

  •  PubChem Substance ID 329751986

Purchase

Properties

InChI Key   ZDRJSYVHDMFHSC-UHFFFAOYSA-M
grade   technical
concentration   ~1 M in THF
density   0.93 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3399 4.3/PG 1
WGK Germany 
1
Flash Point(F) 
1 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

364649 tert-Butylmagnesium chloride solution, 1.0 M in THF

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?