• USA Home
  • 20235 - S-tert-Butylmercapto-L-cysteine

EMAIL THIS PAGE TO A FRIEND
20235 Sigma-Aldrich

S-tert-Butylmercapto-L-cysteine

purum, ≥95.0% (T)

Synonym: 3-(tert-Butyldithio)-L-alanine, S-(tert-Butylthio)-L-cysteine

  • CAS Number 30044-51-0

  • Empirical Formula (Hill Notation) C7H15NO2S2

  • Molecular Weight 209.33

  •  Beilstein Registry Number 2247500

  •  EC Number 250-012-7

  •  MDL number MFCD00002609

  •  PubChem Substance ID 329751995

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Cysteine/Cystine, Peptide Chemistry More...
InChI Key   TWMBHZTWEDJDRC-YFKPBYRVSA-N
grade   purum
assay   ≥95.0% (T)
optical activity   [α]20/D −83±3°, c = 1% in 1 M HCl
impurities   ≤0.5% free cysteine
ign. residue   ≤0.05%
mp   ~190 °C (dec.)

Description

Other Notes

Important S-protected cysteine-derivative for peptide synthesis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B5890 S-tert-Butylmercapto-L-cysteine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?