• USA Home
  • 20247 - tert-Butyl methyl ether

EMAIL THIS PAGE TO A FRIEND
20247 Sigma-Aldrich

tert-Butyl methyl ether

for HPLC, ≥99.8% (GC)

Synonym: MTBE, Methyl tert-butyl ether

  • CAS Number 1634-04-4

  • Linear Formula (CH3)3COCH3

  • Molecular Weight 88.15

  •  Beilstein Registry Number 1730942

  •  EC Number 216-653-1

  •  MDL number MFCD00008812

Purchase

Properties

grade   for HPLC
vapor density   3.1 (vs air)
InChI Key   BZLVMXJERCGZMT-UHFFFAOYSA-N
assay   ≥99.8% (GC)
autoignition temp.   705 °F
shelf life   limited shelf life, expiry date on the label
expl. lim.   15.1 %
impurities   ≤0.05% water
evapn. residue   ≤0.001%
refractive index   n20/D 1.369(lit.)
  n20/D 1.369
bp   55-56 °C(lit.)
density   0.74 g/mL at 25 °C(lit.)
λ   1 cm path, H2O reference
UV absorption   λ: 220 nm Amax: 1.0
  λ: 250 nm Amax: 0.1
  λ: 280 nm Amax: 0.01

Description

Other Notes

A new chromatography eluent

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
1
RTECS 
KN5250000
Flash Point(F) 
-25.6 °F
Flash Point(C) 
-32 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?