• USA Home
  • 20310 - 2-tert-Butyl-6-methyl-phenol

EMAIL THIS PAGE TO A FRIEND
20310 Sigma-Aldrich

2-tert-Butyl-6-methyl-phenol

purum, ≥98.0% (GC)

Synonym: 6-tert-Butyl-o-cresol

  • CAS Number 2219-82-1

  • Linear Formula (CH3)3CC6H3(CH3)OH

  • Molecular Weight 164.24

  •  Beilstein Registry Number 1862362

  •  EC Number 218-734-7

  •  MDL number MFCD00002239

Purchase

Properties

Related Categories Building Blocks, C9 to C20+, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   BKZXZGWHTRCFPX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.519(lit.)
bp   230 °C(lit.)
mp   24-27 °C(lit.)
density   0.967 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2430 8 / PGII
WGK Germany 
3
RTECS 
SK1578200
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?