EMAIL THIS PAGE TO A FRIEND
20520 Sigma-Aldrich

4-tert-Butylphenol

puriss., ≥99.0% (GC)

  • CAS Number 98-54-4

  • Linear Formula (CH3)3CC6H4OH

  • Molecular Weight 150.22

  •  Beilstein Registry Number 1817334

  •  EC Number 202-679-0

  •  MDL number MFCD00002367

Purchase

Properties

Related Categories Building Blocks, C9 to C20+, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
vapor pressure   1 mmHg ( 70 °C)
InChI Key   QHPQWRBYOIRBIT-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
bp   236-238 °C(lit.)
mp   96-101 °C(lit.)
  98-101 °C
solubility   alcohol: soluble
  cold water: very slightly soluble
  diethyl ether: soluble
density   0.908 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

4-tert-Butylphenol was used as a model pollutant to study the degaradation of 4-tert-butylphenol in various conditions.

General description

The effect of 4-tert-butylphenol (BP) in a murine model of atopic dermatitis was studied.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
SJ8925000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?